R&D Systems
Manufacturer Code:FAB16861C100UG
Catalog # FB16861C100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 753317 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R Gly24-Asp372 Accession No. Q8K4B4 |
Conjugate | PerCP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Rat Monoclonal Antibody
Protein Aliases: IBD17 IL-23 R IL-23 receptor IL23R IL-23R interleukin 23 receptor interleukin-23 receptor; IL-23 receptor; Interleukin-23 receptor
Gene Aliases: IL23R
UniProt ID: (Human) Q5VWK5
Entrez Gene ID: (Human) 149233
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.