R&D Systems
Manufacturer Code:IC17161G
Catalog # IC17161G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 727753 |
Immunogen | <EM>E. coli</EM>-derived recombinant human IL-23 p19 Arg20-Pro189 Accession # Q9NPF7 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IL-23 subunit alpha; IL-23-A; IL-23p19; IL23 IL-23 IL-23 p19/IL-12 p40 IL23A IL-23A IL-23-A IL-23p19 IL-23p19/IL-12p40 IL23P19P19 interleukin 23 p19 subunit interleukin 23 alpha subunit p19 interleukin-23 subunit alpha Interleukin-23 subunit p19 JKA3 induced upon T-cell activation MGC79388 SGRF SGRFIL-23 subunit alpha; interleukin 23 p19 subunit; interleukin 23, alpha subunit p19; Interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation
Gene Aliases: IL-23; IL-23A; IL23A; IL23P19; P19; SGRF; UNQ2498/PRO5798
UniProt ID: (Human) Q9NPF7
Entrez Gene ID: (Human) 51561
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.