Novus Biologicals
Manufacturer Code:NBP176697
Catalog # NBP176697
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human IL-23. The immunogen is located within amino acids 40 - 90 of IL-23. Amino Acid Squence: REEGDEETTNDVPHIQC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IL-23 IL-23A IL-23-A IL-23p19 IL23P19P19 interleukin 23 p19 subunit interleukin 23 alpha subunit p19 interleukin-23 subunit alpha Interleukin-23 subunit p19 JKA3 induced upon T-cell activation MGC79388 SGRFIL-23 subunit alpha; IL-23 subunit alpha; IL-23-A; IL-23p19; interleukin 23 p19 subunit; interleukin 23, alpha subunit p19; Interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation
Gene Aliases: IL-23; IL-23A; IL23A; IL23P19; P19; SGRF; UNQ2498/PRO5798
UniProt ID: (Human) Q9NPF7
Entrez Gene ID: (Human) 51561
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.