Invitrogen
Manufacturer Code:50702382
Catalog # 50-112-8877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | fc23cpg |
Conjugate | eFluor 660 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: IL-23 IL-23A IL23P19 MGC79388 P19 SGRF; IL-23 subunit alpha; IL-23-A; IL-23p19; Interleukin-23 subunit alpha; Interleukin-23 subunit p19
Gene Aliases: IL-23; Il23a; p19
UniProt ID: (Mouse) Q9EQ14
Entrez Gene ID: (Mouse) 83430
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.