Invitrogen
Manufacturer Code:165236025
Catalog # 16-523-6025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Functional Assay (FN) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HNU2319 |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: il 23 IL 23 A IL12B Il-12b Il12p40 Il-12p40 il23 IL-23 IL-23 subunit alpha Il23a IL-23A IL-23-A IL23P19 IL-23p19 ILN Interleukin interleukin 12B interleukin 23 p19 subunit interleukin 23 subunit alpha Interleukin 23 alpha subunit p19 interleukin-23 subunit alpha Interleukin-23 subunit p19 interleukin-six G-CSF related factor JKA3 induced upon T-cell activation MGC79388 P19 p40 RP23-388G23.1 SGRF UNQ2498/PRO5798; IL-23 subunit alpha; IL-23-A; IL-23p19; interleukin 23 p19 subunit; interleukin 23, alpha subunit p19; Interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation
Gene Aliases: IL-23; IL-23A; IL23A; IL23P19; P19; SGRF; UNQ2498/PRO5798
UniProt ID: (Human) Q9NPF7
Entrez Gene ID: (Human) 51561
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.