R&D Systems
Manufacturer Code:FAB42941G100UG
Catalog # FA42941G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 496514 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IL-22 R alpha 1 Thr18-Ala228 Accession # Q80XZ4 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Rat MonoclonalAntibody
For Research Use Only
Protein Aliases: CRF2-9; CRF2-9 CRF2-9interleukin 22 receptor Cytokine receptor class-II member 9 Cytokine receptor family 2 member 9 IL-22 receptor subunit alpha-1 IL22R IL22R alpha 1 IL22R1 IL22RA1 IL-22RA1 IL-22R-alpha-1 IL-TIF-R1 interleukin 22 receptor alpha 1 interleukin-22 receptor subunit alpha-1 zcytoR11; Cytokine receptor class-II member 9; Cytokine receptor family 2 member 9; IL-22 receptor subunit alpha-1; IL-22R-alpha-1; IL-22RA1; interleukin 22 receptor, alpha 1; Interleukin-22 receptor subunit alpha-1; ZcytoR11
Gene Aliases: CRF2-9; IL22R; IL22R1; IL22RA1
UniProt ID: (Human) Q8N6P7
Entrez Gene ID: (Human) 58985
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.