Invitrogen
Manufacturer Code:MA524205
Catalog # PIMA524205
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 496514 |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse IL-22 R alpha 1 Thr18-Ala228 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CRF2-9 IL22R IL22R1 IL-22 receptor subunit alpha-1 IL-22R-alpha-1 IL-22RA1 cytokine receptor class-II member 9 cytokine receptor family 2 member 9 interleukin-22 receptor subunit alpha-1 zcytoR11; IL-22 receptor subunit alpha-1; IL-22R-alpha-1; IL-22RA1; interleukin-22 receptor alpha chain; Interleukin-22 receptor subunit alpha-1
Gene Aliases: 9130219A07Rik; ENSMUSG00000073746; IL-22R; Il22r; Il22ra1
UniProt ID: (Mouse) Q80XZ4
Entrez Gene ID: (Mouse) 230828
Molecular Function: cytokine receptor defense/immunity protein receptor type I cytokine receptor type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.