Invitrogen
Manufacturer Code:PA5115409
Catalog # PIPA5115409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IL22 Receptor Alpha corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9130219A07Rik CRF2-9 Cytokine receptor class-II member 9 cytokine receptor family 2 member 9 ENSMUSG00000073746 il-22 receptor IL-22 receptor subunit alpha-1 Il22r IL-22R IL22R1 Il22ra1 IL-22RA1 IL-22R-alpha-1 ILN Interleukin interleukin 22 receptor subunit alpha 1 interleukin 22 receptor alpha 1 interleukin-22 receptor alpha chain Interleukin-22 receptor subunit alpha-1 RGD1559655 ZcytoR11; CRF2-9; Cytokine receptor class-II member 9; Cytokine receptor family 2 member 9; IL-22 receptor subunit alpha-1; IL-22R-alpha-1; IL-22RA1; interleukin 22 receptor, alpha 1; interleukin-22 receptor alpha chain; Interleukin-22 receptor subunit alpha-1; ZcytoR11
Gene Aliases: 9130219A07Rik; CRF2-9; ENSMUSG00000073746; IL-22R; IL22R; IL22R1; IL22RA1; RGD1559655
UniProt ID: (Human) Q8N6P7, (Mouse) Q80XZ4
Entrez Gene ID: (Human) 58985, (Mouse) 230828, (Rat) 362629
Molecular Function: cytokine receptor defense/immunity protein receptor type I cytokine receptor type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.