R&D Systems
Manufacturer Code:FAB9911P025
Catalog # FAB9911P025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 152512 |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human IL-21 R Cys20-Pro236 Accession # Q9HBE5 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD360; CD360 CD360 antigen IL-21 R IL-21 receptor IL21R IL-21R interleukin 21 receptor interleukin-21 receptor MGC10967 NILR Novel interleukin receptor; IL-21 receptor; Interleukin-21 receptor; Novel interleukin receptor
Gene Aliases: CD360; IL21R; NILR; UNQ3121/PRO10273
UniProt ID: (Human) Q9HBE5
Entrez Gene ID: (Human) 50615
Molecular Function:
cytokine receptor
receptor
type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.