Novus Biologicals
Manufacturer Code:NBP289468UV
Catalog # NB107791
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 24 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human IL-20R alpha (NP_055247.3 Met 1-Lys 250). |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: class II cytokine receptor ZCYTOR7; class II cytokine receptor ZCYTOR7 CRF2-8 Cytokine receptor class-II member 8 Cytokine receptor family 2 member 8 FLJ40993 IL-20R1IL-20 receptor subunit alpha IL-20RA IL-20R-alpha interleukin 20 receptor alpha interleukin-20 receptor I interleukin-20 receptor subunit alpha ZcytoR7; CRF2-8; Cytokine receptor class-II member 8; Cytokine receptor family 2 member 8; IL-20 receptor subunit alpha; IL-20R1; interleukin 20 receptor alpha subunit; interleukin 20 receptor, alpha; interleukin-20 receptor I; Interleukin-20 receptor subunit alpha; ZcytoR7
Gene Aliases: CRF2-8; IL-20R-alpha; IL-20R1; IL-20RA; IL20RA; UNQ681/PRO1315
UniProt ID: (Human) Q9UHF4
Entrez Gene ID: (Human) 53832
Molecular Function: cytokine receptor defense/immunity protein receptor type I cytokine receptor type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.