R&D Systems
Manufacturer Code:MAB5891
Catalog # MAB5891
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 130122 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IL-2 R beta Ala27-Glu240 Accession # P16297 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD122; CD122 antigen; CD122 CD122 antigen high affinity IL-2 receptor beta subunit High affinity IL-2 receptor subunit beta IL-15 R beta IL-2 receptor subunit beta IL2R beta IL-2R subunit beta IL2RB IL-2RB interleukin 15 receptor beta interleukin 2 receptor beta interleukin-2 receptor subunit beta P70-75 p75; high affinity IL-2 receptor beta subunit; High affinity IL-2 receptor subunit beta; IL-2 receptor subunit beta; IL-2R subunit beta; IL-2RB; interleukin 15 receptor, beta; interleukin 2 receptor, beta; Interleukin-15 receptor subunit beta; Interleukin-2 receptor subunit beta; p70-75; p75
Gene Aliases: CD122; IL15RB; IL2RB; P70-75
UniProt ID: (Human) P14784
Entrez Gene ID: (Human) 3560
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.