R&D Systems
Manufacturer Code:AF502NA
Catalog # AF502NA
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant rat IL-2 Ala21-Gln155 Accession # P17108 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Aldesleukin; aldesleukin IL2 IL-2lymphokine interleukin 2 interleukin-2 involved in regulation of T-cell clonal expansion T cell growth factor T-cell growth factor TCGF; IL-2; Interleukin-2; involved in regulation of T-cell clonal expansion; T cell growth factor; T-cell growth factor; TCGF
Gene Aliases: IL-2; IL2; lymphokine; TCGF
UniProt ID: (Human) P60568
Entrez Gene ID: (Human) 3558
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.