Novus Biologicals
Manufacturer Code:NBP211678AF647
Catalog # NB211678647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0379-10G36 |
Immunogen | Produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-18 binging protein A (IL-18 BPa). |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: IL-18BP; IL-18BP IL18BPa interleukin 18 binding protein interleukin-18-binding protein MC51L-53L-54L homolog gene product tadekinig-alfa; Interleukin-18-binding protein; MC51L-53L-54L homolog gene product; Tadekinig-alfa
Gene Aliases: IL18BP; IL18BPa
UniProt ID: (Human) O95998
Entrez Gene ID: (Human) 10068
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.