Invitrogen
Manufacturer Code:MA530766
Catalog # PIMA530766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 169 |
Immunogen | Recombit Mouse IL18/IL-18/Interleukin 18 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: IFN-gamma-inducing factor; IL-1 gamma; IL-18; IL18 interleukin 18 (interferon-gamma-inducing factor) IGIF IL-18 IL-1g IL1F4 MGC12320 IL-1 gamma interferon-gamma-inducing factor interleukin 18 interleukin-1 gamma interleukin-18; Interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin-1 gamma; Interleukin-18
Gene Aliases: Igif; Il-18; Il18
UniProt ID: (Mouse) P70380
Entrez Gene ID: (Mouse) 16173
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.