Invitrogen
Manufacturer Code:PA579477
Catalog # PIPA579477
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of Human IL18. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Iboctadekin; IFN-gamma-inducing factor; IL-1 gamma; IL-18; Interferon gamma-inducing factor; interleukin 18 (interferon-gamma-inducing factor); Interleukin-1 gamma; Interleukin-18; Interleukin-18 IL-18 Iboctadekin Interferon gamma-inducing factor IFN-gamma-inducing factor Interleukin-1 gamma IL-1 gamma IL18 IGIF IL1F4
Gene Aliases: IGIF; IL-18; IL-1g; IL18; IL1F4
UniProt ID: (Human) Q14116
Entrez Gene ID: (Human) 3606
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.