Invitrogen
Manufacturer Code:PA547344
Catalog # PIPA547344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit rhesus macaque IL-18/IL-1F4 Tyr37-Asp193 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: cytokine; IL18 interleukin-18 interferon-gamma-inducing factor IGIF IL-1g IL1F4 IL-1 gamma interferon-gamma-inducing factor interleukin-1 gamma; interleukin 18 (interferon-gamma-inducing factor); interleukin-18 proprotein
Entrez Gene ID: (Rhesus monkey) 574151
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.