Novus Biologicals
Manufacturer Code:NBP289540100UL
Catalog # NB020563
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | Recombinant Human IL-17RD/SEF Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ35755 IL-17RDIL-17 receptor D IL17Rhom IL17RLMDKFZp434N1928 interleukin 17 receptor D interleukin-17 receptor D Interleukin-17 receptor-like protein MGC133309 Sef homolog SEFhSef; hSef; IL-17 receptor D; IL17Rhom; Interleukin-17 receptor D; Interleukin-17 receptor-like protein; Sef homolog
Gene Aliases: HH18; IL-17RD; IL17RD; IL17RLM; SEF; UNQ6115/PRO20026
UniProt ID: (Human) Q8NFM7
Entrez Gene ID: (Human) 54756
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.