R&D Systems
Manufacturer Code:AF2276SP
Catalog # AF2276SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RD/SEF Gly28-Arg299 Accession # Q8JZL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ35755 IL-17 RD IL17RD IL-17RDIL-17 receptor D IL17Rhom IL17RLM IL17RLMDKFZp434N1928 interleukin 17 receptor D interleukin-17 receptor D Interleukin-17 receptor-like protein MGC133309 SEF Sef homolog SEFhSef; hSef; IL-17 receptor D; IL17Rhom; Interleukin-17 receptor D; Interleukin-17 receptor-like protein; Sef homolog
Gene Aliases: HH18; IL-17RD; IL17RD; IL17RLM; SEF; UNQ6115/PRO20026
UniProt ID: (Human) Q8NFM7
Entrez Gene ID: (Human) 54756
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.