Novus Biologicals
Manufacturer Code:NBP185450
Catalog # NBP185450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:QCGSETGPSPEWMLQHDLIPGDLRDLRVEPVTTSVATGDYSILMNVSWVLRADASIRLLKATKICVTGKSNFQSYSCVRCNYTEAFQTQTRPSGGKWTFSYIGFPVELNTVYFIGAHNIPNANMNEDGPSMSVNFTSPG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytokine Zcyto7; Cytokine Zcyto7 cytokine-like protein ZCYTO7 IL-17Binterleukin-17 beta IL20 IL-20interleukin-17B interleukin 17B interleukin 20 Interleukin-20 MGC138900 MGC138901 neuronal interleukin-17 related factor Neuronal interleukin-17-related factor NIRF ZCYTO7interleukin-20; cytokine-like protein ZCYTO7; IL-17B; IL-20; interleukin 20; interleukin-17 beta; Interleukin-17B; Interleukin-20; neuronal interleukin-17 related factor; Neuronal interleukin-17-related factor
Gene Aliases: IL-17B; IL-20; IL17B; IL20; NIRF; UNQ516/PRO1031; ZCYTO7
UniProt ID: (Human) Q9UHF5
Entrez Gene ID: (Human) 27190
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.