R&D Systems
Manufacturer Code:AF1207SP
Catalog # AF1207SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human IL-17 RB Arg18-Gly289 Accession # NP_061195 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CRL4IL-17 receptor homolog 1 cytokine receptor CRL4 Cytokine receptor-like 4 Evi27 EVI27IL-17B receptor IL-17 RB IL-17 receptor B IL-17 RH1 IL17BRinterleukin 17B receptor IL-17ER IL17RB IL-17RB IL17Rh1 IL-17Rh1 IL17RH1MGC5245 interleukin 17 receptor B interleukin 17 receptor homolog 1 interleukin-17 receptor B Interleukin-17B receptor; cytokine receptor CRL4; Cytokine receptor-like 4; IL-17 receptor B; IL-17 receptor homolog 1; IL-17B receptor; IL-17RB; IL-17Rh1; interleukin 17 receptor homolog 1; interleukin 17B receptor; Interleukin-17 receptor B; Interleukin-17B receptor
Gene Aliases: CRL4; EVI27; IL17BR; IL17RB; IL17RH1; UNQ2501/PRO19612
UniProt ID: (Human) Q9NRM6
Entrez Gene ID: (Human) 55540
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.