R&D Systems
Manufacturer Code:AF448
Catalog # AF448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant mouse IL-17 RA/IL-17 R Extracellular domain |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD217; CD217 CD217 antigen Cdw217 CDw217interleukin 17 receptor hIL-17R IL-17 R IL-17 RA IL-17 receptor A IL17RA IL-17RAMGC10262 IL17Rinterleukin-17 receptor A interleukin 17 receptor A; CDw217; IL-17 receptor A; Interleukin-17 receptor A
Gene Aliases: CANDF5; CD217; CDw217; hIL-17R; IL-17RA; IL17R; IL17RA
UniProt ID: (Human) Q96F46
Entrez Gene ID: (Human) 23765
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.