Invitrogen
Manufacturer Code:167178050
Catalog # 16-717-8050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | eBio64CAP17 |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CTLA-8; CTLA8 Ctla-8 cytokine cytotoxic T-lymphocyte-associated antigen 8 cytotoxic T-lymphocyte-associated protein 8 il 17 Il17 IL-17 Il17a IL-17A ILN Interleukin interleukin 17 Interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8) interleukin 17 precursor interleukin 17A interleukin 17A-like interleukin-17 Interleukin17A interleukin-17A LOC100339322 R-IL-17-A; Cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; IL-17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); Interleukin-17A
Gene Aliases: CTLA-8; CTLA8; IL-17; IL-17A; IL17; IL17A
UniProt ID: (Human) Q16552
Entrez Gene ID: (Human) 3605
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.