Invitrogen
Manufacturer Code:48717782
Catalog # 50-112-4211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | eBio17B7 |
Conjugate | eFluor 450 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CTLA-8; Cytotoxic T-lymphocyte-associated antigen 8; IL-17; Interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); Interleukin-17; Interleukin-17A
Gene Aliases: CTLA-8; Ctla8; IL-17; IL-17A; Il17; Il17a
UniProt ID: (Mouse) Q62386
Entrez Gene ID: (Mouse) 16171, (Rat) 301289
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.