Invitrogen
Manufacturer Code:PA546947
Catalog # PIPA546947
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse IL-17 Thr22-Ala158 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CTLA-8; CTLA8 IL17 interleukin 17 cytotoxic T-lymphocyte-associated serine esterase 8 Interleukin-17A; Cytotoxic T-lymphocyte-associated antigen 8; IL-17; Interleukin-17A
Gene Aliases: Ctla-8; Ctla8; IL-17; IL-17A; Il17; Il17a
UniProt ID: (Mouse) Q62386
Entrez Gene ID: (Mouse) 16171
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.