R&D Systems
Manufacturer Code:MAB317100
Catalog # MAB317100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 41809 |
Immunogen | <EM>E. coli</EM>-derived recombinant human IL-17 Ile20-Ala155 Accession # Q16552 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CTLA-8; CTLA8 CTLA-8 CTLA8cytotoxic T-lymphocyte-associated serine esterase 8 Cytotoxic T-lymphocyte-associated antigen 8 IL17 IL17A IL-17A IL-17Acytotoxic T-lymphocyte-associated protein 8 IL-17CTLA-8 IL17interleukin-17A interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8) interleukin 17A; Cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; IL-17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); Interleukin-17A
Gene Aliases: CTLA-8; CTLA8; IL-17; IL-17A; IL17; IL17A
UniProt ID: (Human) Q16552
Entrez Gene ID: (Human) 3605
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.