Novus Biologicals
Manufacturer Code:NBP176337
Catalog # NBP176337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 19 amino acid synthetic peptide from near the center of human IL-17A. The immunogen is located within amino acids 50 - 100 of IL-17. Amino Acid Squence: YYNRSTSPWNLHRNEDPER |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTLA-8; CTLA8cytotoxic T-lymphocyte-associated serine esterase 8 Cytotoxic T-lymphocyte-associated antigen 8 IL-17Acytotoxic T-lymphocyte-associated protein 8 IL-17CTLA-8 IL17interleukin-17A interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8) interleukin 17A; Cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; IL-17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); Interleukin-17A
Gene Aliases: CTLA-8; CTLA8; IL-17; IL-17A; IL17; IL17A
UniProt ID: (Human) Q16552
Entrez Gene ID: (Human) 3605
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.