Proteintech
Manufacturer Code:661481IG150UL
Catalog # 50-173-6582
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B3D5 |
Immunogen | IL-17 Fusion Protein Ag3733 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CTLA 8 CTLA8 IL 17 IL 17A IL17 IL-17 IL17A interleukin 17A; CTLA-8; Cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; IL-17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); Interleukin-17A
Gene Aliases: CTLA-8; CTLA8; IL-17; IL-17A; IL17; IL17A
UniProt ID: (Human) Q16552
Entrez Gene ID: (Human) 3605
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.