R&D Systems
Manufacturer Code:AF539SP
Catalog # AF539SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IL-13 R alpha 2 Leu22-Lys334 Accession # O88786 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer/testis antigen 19; cancer/testis antigen 19 CD213a2 CD213a2 antigen CT19 IL-13 receptor subunit alpha-2 IL13BP IL13R IL-13R IL13R alpha 2 IL-13R subunit alpha-2 IL13RA2 IL-13RA2 IL-13R-alpha-2 interleukin 13 binding protein interleukin 13 receptor alpha 2 chain interleukin 13 receptor alpha 2 interleukin-13 receptor subunit alpha-2 Interleukin-13-binding protein; CD213a2; IL-13 receptor subunit alpha-2; IL-13R subunit alpha-2; IL-13R-alpha-2; IL-13RA2; interleukin 13 binding protein; interleukin 13 receptor alpha 2 chain; interleukin 13 receptor, alpha 2; Interleukin-13 receptor subunit alpha-2; Interleukin-13-binding protein
Gene Aliases: CD213A2; CT19; IL-13R; IL13BP; IL13R; IL13RA2
UniProt ID: (Human) Q14627
Entrez Gene ID: (Human) 3598
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.