Invitrogen
Manufacturer Code:167135050
Catalog # 16-713-5050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | eBio1316H |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ALRH ALRH antibody BHR1 il 13 Il13 Il-13 IL-13 cytokine ILN Interleukin interleukin 13 Interleukin13 Interleukin-13 interleukin-13 precursor L13 MGC116786 MGC116788 MGC116789 NC30 NC30 antibody P600 P600 antibody T-cell activation protein P600; IL-13; Interleukin-13; T-cell activation protein P600
Gene Aliases: Il-13; Il13
UniProt ID: (Mouse) P20109
Entrez Gene ID: (Mouse) 16163
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.