R&D Systems
Manufacturer Code:MAB213100
Catalog # MAB213100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Neutralization (Neu) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 32116 |
Immunogen | <EM>E. coli</EM>-derived recombinant human IL-13 Gly35-Asn146 Accession # P35225 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ALRHMGC116789 BHR1interleukin-13 IL13 IL-13MGC116788 interleukin 13 MGC116786 NC30 P600; IL-13; Interleukin-13
Gene Aliases: IL-13; IL13; NC30; P600
UniProt ID: (Human) P35225
Entrez Gene ID: (Human) 3596
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.