R&D Systems
Manufacturer Code:MAB219500
Catalog # MAB219500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 24910 |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human IL-12 p70 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CLMF p35; CLMF p35 CLMF1 Cytotoxic lymphocyte maturation factor 35 kDa subunit IL12 IL12 p70 IL-12 subunit p35 IL-12 subunit p35 IL12A interleukin 12 p35 interleukin 12A (natural killer cell stimulatory factor 1 cytotoxic lymphocytematuration factor 1 p35) interleukin-12 alpha chain interleukin-12 subunit alpha natural killer cell stimulatory factor 1 35 kD subunit NF cell stimulatory factor chain 1 NK cell stimulatory factor chain 1 NKSF1 p35; cytotoxic lymphocyte maturation factor 1, p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; IL-12 subunit p35; IL-12, subunit p35; IL-12A; IL35 subunit; interleukin 12, p35; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin-12 alpha chain; Interleukin-12 subunit alpha; NF cell stimulatory factor chain 1; NK cell stimulatory factor chain 1; NKSF1
Gene Aliases: CLMF; IL-12A; IL12A; NFSK; NKSF1; P35
UniProt ID: (Human) P29459
Entrez Gene ID: (Human) 3592
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.