Invitrogen
Manufacturer Code:PA547961
Catalog # PIPA547961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Pig |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO-derived recombit porcine IL-12 p35/p40 heterodimer Ile23-Asn324 (p40) Arg26-Ser222 (p35) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CLMF p35; CLMF p40; Cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; IL-12 subunit p35; IL-12 subunit p40; IL-12A; IL-12B; IL12A IL12B Interleukin-12 subunit alpha IL-12A Cytotoxic lymphocyte maturation factor 35 kDa subunit CLMF p35 IL-12 subunit p35 NK cell stimulatory factor chain 1 NKSF1 NKSF1 Interleukin-12 subunit beta IL-12B Cytotoxic lymphocyte maturation factor 40 kDa subunit IL-12 subunit p40 NK cell stimulatory factor chain 2 NKSF2 NKSF2 IL12p35 IL12P40; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin-12 p35 subunit; interleukin-12 p40 subunit; Interleukin-12 subunit alpha; Interleukin-12 subunit beta; interleukin-23 p40 subunit
Gene Aliases: IL12A; IL12B
UniProt ID: (Pig) Q29053, (Pig) Q28938
Entrez Gene ID: (Pig) 397053, (Pig) 397076
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.