Invitrogen
Manufacturer Code:PA594920
Catalog # PIPA594920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived rat IL-12 (p40) recombit protein (Position: M23-S321). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: interleukin-12 p40; interleukin-12 subunit beta; Interleukin-12 subunit beta Cytotoxic lymphocyte maturation factor 40 kDa subunit IL-12 subunit p40 Il12b IL12B
Gene Aliases: Il12
Entrez Gene ID: (Rat) 64546
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.