Invitrogen
Manufacturer Code:PA547197
Catalog # PIPA547197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Sf 21-derived recombit rat IL-12 p40: Met23-Ser335 p35: Arg23-Ser215 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: IL12A IL12B Interleukin-12 subunit alpha IL-12A Cytotoxic lymphocyte maturation factor 35 kDa subunit CLMF p35 IL-12 subunit p35 NK cell stimulatory factor chain 1 NKSF1 NKSF1 Interleukin-12 subunit beta IL-12B Cytotoxic lymphocyte maturation factor 40 kDa subunit IL-12 subunit p40 NK cell stimulatory factor chain 2 NKSF2 NKSF2; interleukin-12 p40; interleukin-12 subunit beta
Gene Aliases: Il12
Entrez Gene ID: (Rat) 64546
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.