Invitrogen
Manufacturer Code:PA5115402
Catalog # PIPA5115402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IL12B corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLMF CLMF p35 CLMF p40 CLMF2 Cytotoxic lymphocyte maturation factor 40 kDa subunit il 12 Il12 IL-12 p40 subunit IL-12 subunit p35 IL-12 subunit p40 IL12 subunit p40 IL-12.p40 IL12A IL-12A Il12b IL-12B Il12p40 Il-12p40 ILN IMD28 IMD29 Interleukin Interleukin 12 interleukin 12 p40 subunit interleukin 12 p40 interleukin 12B interleukin 12B (natural killer cell stimulatory factor 2 cytotoxic lymphocyte maturation factor 2 p40) Interleukin12 interleukin-12 beta chain interleukin-12 p40 interleukin-12 p40 chain interleukin-12 p40 subunit interleukin-12 subunit beta interleukin-12p40 interleukin-23 p40 subunit natural killer cell stimulatory factor 40 kD subunit NK cell stimulatory factor chain 2 NKSF NKSF1 NKSF2 p40 RP23-388G23.1; CLMF p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit; IL-12 subunit p40; IL-12B; IL12, subunit p40; interleukin 12, p40; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin-12 beta chain; interleukin-12 p40; Interleukin-12 subunit beta; natural killer cell stimulatory factor, 40 kD subunit; NK cell stimulatory factor chain 2; NKSF2
Gene Aliases: CLMF; CLMF2; IL-12B; Il-12p40; Il12; IL12B; Il12p40; IMD28; IMD29; NKSF; NKSF2; p40
UniProt ID: (Human) P29460, (Mouse) P43432
Entrez Gene ID: (Human) 3593, (Rat) 64546, (Mouse) 16160
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.