Invitrogen
Manufacturer Code:14712882
Catalog # 50-112-8691
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | B-T21 (BT21) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CLMF IL-12A NFSK NKSF1 P35 IL12p35; CLMF p35; cytotoxic lymphocyte maturation factor 1, p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; IL-12 subunit p35; IL-12, subunit p35; IL-12A; IL35 subunit; interleukin 12, p35; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin-12 alpha chain; Interleukin-12 subunit alpha; NF cell stimulatory factor chain 1; NK cell stimulatory factor chain 1; NKSF1
Gene Aliases: CLMF; IL-12A; IL12A; NFSK; NKSF1; P35
UniProt ID: (Human) P29459
Entrez Gene ID: (Human) 3592
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.