Novus Biologicals
Manufacturer Code:NBP211659AF594
Catalog # NP21169A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0363-7V24 |
Immunogen | produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-12R b1 extracellular domain. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CD212; CD212 CD212 antigen IL-12 receptor beta component IL-12 receptor subunit beta-1 IL12R IL-12R subunit beta-1 IL12RB IL-12RB1 IL-12R-beta-1 IL-12R-BETA1 interleukin 12 receptor beta 1 interleukin-12 receptor beta-1 chain interleukin-12 receptor subunit beta-1 MGC34454; cluster of differentiation 212; IL-12 receptor beta component; IL-12 receptor subunit beta-1; IL-12R subunit beta-1; interleukin 12 receptor, beta 1; interleukin-12 receptor beta-1 chain; Interleukin-12 receptor subunit beta-1
Gene Aliases: CD212; IL-12R-BETA1; IL12R; IL12RB; IL12RB1; IMD30
UniProt ID: (Human) P42701
Entrez Gene ID: (Human) 3594
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.