Invitrogen
Manufacturer Code:12712341
Catalog # 50-111-52
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | C17.8 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CLMF p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit; IL-12 subunit p40; IL-12B; IL12B Il12p40 interleukin 12B RP23-388G23.1 Il-12b Il-12p40 p40 IL-12 p40 IL-23 subunit p40 IL12P40; Interleukin-12 subunit beta
Gene Aliases: Il-12b; Il-12p40; Il12b; Il12p40; p40
UniProt ID: (Mouse) P43432
Entrez Gene ID: (Mouse) 16160
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.