R&D Systems
Manufacturer Code:MAB4992100
Catalog # MAB4992100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2329A |
Immunogen | <em>S. frugiperda</em> <em>Sf 21</em>-derived recombinant mouse IL-12/IL-23 p40 protein Met1-Ser135 Accession # P43432 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Rabbit Monoclonal Antibody
Protein Aliases: CLMF CLMF p40 CLMF2 Cytotoxic lymphocyte maturation factor 40 kDa subunit IL12 p40 IL-12 subunit p40 IL12B IL-12B IL-12BNK cell stimulatory factor chain 2 interleukin 12 p40 interleukin 12B (natural killer cell stimulatory factor 2 cytotoxic lymphocytematuration factor 2 p40) interleukin-12 beta chain interleukin-12 subunit beta natural killer cell stimulatory factor 40 kD subunit NKSF NKSF2 NKSF2IL12 subunit p40; CLMF p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit; IL-12 subunit p40; IL-12B; IL12, subunit p40; interleukin 12, p40; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin-12 beta chain; Interleukin-12 subunit beta; natural killer cell stimulatory factor, 40 kD subunit; NK cell stimulatory factor chain 2; NKSF2
Gene Aliases: CLMF; CLMF2; IL-12B; IL12B; IMD28; IMD29; NKSF; NKSF2
UniProt ID: (Human) P29460
Entrez Gene ID: (Human) 3593
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.