Invitrogen
Manufacturer Code:MA530732
Catalog # PIMA530732
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 101 |
Immunogen | Recombit Mouse IL-23 (IL23A & IL12B Heterodimer) Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CLMF p35 IL-12 subunit p35 IL-12 subunit p35 NF cell stimulatory factor chain 1 NK cell stimulatory factor chain 1 cytotoxic lymphocyte maturation factor 1 p35 cytotoxic lymphocyte maturation factor 35 kDa subunit interleukin 12 p35 interleukin-12 alpha chain interleukin-12 subunit alpha natural killer cell stimulatory factor 1 35 kD subunit IL12p35 IL12P40; CLMF p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit; IL-12 subunit p40; IL-12B; Interleukin-12 subunit beta
Gene Aliases: Il-12b; Il-12p40; Il12b; Il12p40; p40
UniProt ID: (Mouse) P43432
Entrez Gene ID: (Mouse) 16160
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.