Invitrogen
Manufacturer Code:MA523777
Catalog # PIMA523777
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Species reactivity | Pig |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 148801 |
Immunogen | E. coli-derived recombit porcine IL-10 Ser19-Asn175 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CSIF; CSIF IL10A MGC126450 MGC126451 TGIF Interleukin-10; Cytokine synthesis inhibitory factor; IL-10; Interleukin-10
Gene Aliases: CSIF; IL-10; IL10
UniProt ID: (Pig) Q29055
Entrez Gene ID: (Pig) 397106
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.