Invitrogen
Manufacturer Code:MP425
Catalog # PIMP425
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Pig |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | DF8 |
Immunogen | Recombit porcine IL-1 beta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IL-1 beta; IL-1 beta interleukin-1 beta prointerleukin-1 beta IL-1beta; interleukin 1, beta 1; Interleukin-1 beta; prointerleukin-1 beta
Gene Aliases: IL1B; IL1B1
UniProt ID: (Pig) P26889
Entrez Gene ID: (Pig) 397122
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.