Invitrogen
Manufacturer Code:MA523721
Catalog # PIMA523721
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 30311 |
Immunogen | E. coli-derived recombit mouse IL-1 beta/IL-1F2 Val118-Ser269 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: IL-1 beta; IL-1 beta catabolin interleukin-1 beta preinterleukin 1 beta pro-interleukin-1-beta IL1B IL1 B IL-1beta; Interleukin-1 beta
Gene Aliases: Il-1b; IL-1beta; Il1b
UniProt ID: (Mouse) P10749
Entrez Gene ID: (Mouse) 16176
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.