Invitrogen
Manufacturer Code:16701138
Catalog # 16-701-138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | ALF-161 |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: CTLA8 Hematopoietin-1 IL 1 a IL 1a IL1 IL-1 alpha IL17 Il1a Il-1a IL-1alpha IL1-ALPHA IL1F1 IL1a ILN Interleukin interleukin 1 alpha interleukin 1 alpha interleukin 17 interleukin 1-alpha interleukin 1-alpha (AA 1 - 152) Interleukin1 alpha interleukin-1 alpha interleukin-1 alpha precursor Interleukin-17A preinterleukin 1 alpha pre-interleukin-1 alpha pro-interleukin-1-alpha RP23-160G19.8; IL-1 alpha; Interleukin-1 alpha
Gene Aliases: Il-1a; Il1a
UniProt ID: (Mouse) P01582
Entrez Gene ID: (Mouse) 16175
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.