Invitrogen
Manufacturer Code:PA547876
Catalog # PIPA547876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Pig |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit porcine IL-1 alpha Gln119-Ser270 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: IL-1 alpha; IL-1 alpha hematopoietin-1 interleukin-1 alpha preinterleukin 1 alpha pro-interleukin-1-alpha Hematopoietin-1 IL1A; Interleukin-1 alpha
Gene Aliases: IL-1alpha; IL1A
UniProt ID: (Pig) P18430
Entrez Gene ID: (Pig) 397094
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.