Invitrogen
Manufacturer Code:PA175178
Catalog # PIPA175178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit human IL-1a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Hematopoietin-1; IL-1 alpha; interleukin 1, alpha; Interleukin-1 alpha; preinterleukin 1 alpha; pro-interleukin-1-alpha; RP23-160G19.8 Il-1a
Gene Aliases: IL-1A; IL1; IL1-ALPHA; IL1A; IL1F1
UniProt ID: (Human) P01583
Entrez Gene ID: (Human) 3552
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.