Novus Biologicals
Manufacturer Code:NBP211646AF647
Catalog # NB211646647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0356-4T28 |
Immunogen | Produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-1 R II extracellular domain. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: antigen CDw121b; beta CD121b CD121b antigen IL1RBCD121 antigen-like family member B IL-1R-beta IL-1RT2 IL-1RT-2 interleukin 1 receptor type II Interleukin-1 receptor beta MGC47725; CD121 antigen-like family member B; CD121b; CDw121b; IL-1 type II receptor; IL-1R-2; IL-1R-beta; interleukin 1 receptor type II variant 3; interleukin 1 receptor, type II; Interleukin-1 receptor beta; Interleukin-1 receptor type 2; Interleukin-1 receptor type 2, membrane form; Interleukin-1 receptor type 2, soluble form; Interleukin-1 receptor type II; mIL-1R2; sIL-1R2; type II interleukin-1 receptor, beta
Gene Aliases: CD121b; CDw121b; IL-1R-2; IL-1RT-2; IL-1RT2; IL1R2; IL1R2c; IL1RB
UniProt ID: (Human) P27930
Entrez Gene ID: (Human) 7850
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.