Novus Biologicals
Manufacturer Code:NBP176464SS
Catalog # NBP176464SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human IL-1RAcP. The immunogen is located within the last 50 amino acids of IL-1RAcP. Amino Acid Squence: KWKGEKSKYPQGRFWK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: C3orf13IL-1RAcPIL1R3Interleukin-1 receptor 3 FLJ37788 IL-1 receptor accessory protein IL-1R3 IL-1R-3 interleukin 1 receptor accessory protein interleukin-1 receptor accessory protein interleukin-1 receptor accessory protein beta; IL-1 receptor accessory protein; IL-1R-3; IL-1R3; Interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta
Gene Aliases: C3orf13; IL-1RAcP; IL1R3; IL1RAP
UniProt ID: (Human) Q9NPH3
Entrez Gene ID: (Human) 3556
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.