Proteintech
Manufacturer Code:120161AP150UL
Catalog # 50-172-7644
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IKZF1 Fusion Protein Ag2631 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLL-associated antigen KW-6; DNA binding protein Ikaros hIk 1 Hs.54452 IK1 IKAROS IKZF1 LYF1 PRO0758 ZNFN1A1; DNA-binding protein Ikaros; IKAROS family zinc finger 1 (Ikaros); Ikaros family zinc finger protein 1; Lymphoid transcription factor LyF-1; protein phosphatase 1, regulatory subunit 92; zinc finger protein, subfamily 1A, 1 (Ikaros)
Gene Aliases: 5832432G11Rik; CVID13; hlk-1; Hs.54452; IK1; IKAROS; IKZF1; LyF-1; LYF1; mKIAA4227; PPP1R92; PRO0758; RGD1562979; Zfpn1a1; ZNFN1A1
UniProt ID: (Human) Q13422
Entrez Gene ID: (Human) 10320, (Mouse) 22778, (Rat) 305501
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.