Novus Biologicals
Manufacturer Code:NBP266904
Catalog # NBP266904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JA11-83 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: FIP-3; FIP-3IP2 FIP3NF-kappa-B essential modifier Fip3p I-kappa-B kinase subunit gamma IkB kinase gamma subunit IkB kinase subunit gamma IkB kinase-associated protein 1 IKKAP1 IKKG IKK-gammaIPD2 incontinentia pigmenti inhibitor of kappa light polypeptide gene enhancer in B-cells kinase gamma Inhibitor of nuclear factor kappa-B kinase subunit gamma IP IP1 NEMOAMCBX1 NFkappaB essential modulator NF-kappa-B essential modulator; I-kappa-B kinase subunit gamma; ikB kinase subunit gamma; IkB kinase-associated protein 1; IKKAP1; incontinentia pigmenti; Inhibitor of nuclear factor kappa-B kinase subunit gamma; NEMO; NF-kappa-B essential modifier; NF-kappa-B essential modulator
Gene Aliases: AMCBX1; FIP-3; FIP3; Fip3p; IKBKG; IKK-gamma; IKKAP1; IKKG; IMD33; IP; IP1; IP2; IPD2; NEMO; ZC2HC9
UniProt ID: (Human) Q9Y6K9
Entrez Gene ID: (Human) 8517
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.